Learn More
Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes
Ensure positive identification from grossing to filing with the three-sided writing surface of Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes.
Manufacturer:  Thermo Scientific™ 58925
 View more versions of this product
Catalog No. 58-925
  Check Availability
Add to cart
Specifications
Description
Ensure positive identification from grossing to filing with the three-sided writing surface of Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes.
  • Hinge design ensures specimen safety during processing and easy removal of cover while embedding
  • Base is compatible with metal lids and fits all popular size base molds (metal and disposable plastic)
  • Cassettes are conveniently available assembled or unassembled to facilitate usage in a labeling system or with metal lids
  • Solvent- and decalcifier-resistant and compatible with microwave techniques
  • Different colors allow for easy specimen coding
Description & Specifications
Specifications
Biopsy Cassettes, Small Square Pore
Aqua
250/box; 2 boxes
Aqua; 2 boxes of 250