Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes

Ensure positive identification from grossing to filing with the three-sided writing surface of Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes.

Manufacturer: Thermo Scientific™ 58930

 View more versions of this product

Catalog No. 22-050-140

  Check Availability
Add to Cart

    Specifications

    Color White
    Description 250/box; 2 boxes
    Closure Type Hinged

    Ensure positive identification from grossing to filing with the three-sided writing surface of Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes.

    • Hinge design ensures specimen safety during processing and easy removal of cover while embedding
    • Base is compatible with metal lids and fits all popular size base molds (metal and disposable plastic)
    • Cassettes are conveniently available assembled or unassembled to facilitate usage in a labeling system or with metal lids
    • Solvent- and decalcifier-resistant and compatible with microwave techniques
    • Different colors allow for easy specimen coding

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit