Learn More
Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes Available on GSA/VA Contract for Federal Government customers only.
Ensure positive identification from grossing to filing with the three-sided writing surface of Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes.
Manufacturer:  Thermo Scientific™ 58933
 View more versions of this product
Catalog No. 22-050-143
  Check Availability
Add to cart
Specifications
Description
Ensure positive identification from grossing to filing with the three-sided writing surface of Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes.
  • Hinge design ensures specimen safety during processing and easy removal of cover while embedding
  • Base is compatible with metal lids and fits all popular size base molds (metal and disposable plastic)
  • Cassettes are conveniently available assembled or unassembled to facilitate usage in a labeling system or with metal lids
  • Solvent- and decalcifier-resistant and compatible with microwave techniques
  • Different colors allow for easy specimen coding
Description & Specifications
Specifications
Yellow
250/box; 2 boxes
250/box; 2 boxes; Yellow; Assembled