Learn More
Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes
Ensure positive identification from grossing to filing with the three-sided writing surface of Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes.
Specifications
Type Biopsy Cassettes, Small Square Pore
View More Specs
Products 7
Catalog Number Mfr. No. Color Description Price Quantity & Availability  
Catalog Number Mfr. No. Color Description Price Quantity & Availability  
22-050-142
View Documents
Thermo Scientific™
58932
Green 250/box; 2 boxes This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
58-925
View Documents
Thermo Scientific™
58925
Aqua 250/box; 2 boxes This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
58-926
View Documents
Thermo Scientific™
58926
Lilac 250/box; 2 boxes This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
22-050-141
View Documents
Thermo Scientific™
58931
Blue 250/box; 2 boxes This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
58-928
View Documents
Thermo Scientific™
58928
Pink 500/Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
58-929
View Documents
Thermo Scientific™
58929
Tan 250/box; 2 boxes This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
22-050-140
View Documents
Thermo Scientific™
58930
White 250/box; 2 boxes This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Ensure positive identification from grossing to filing with the three-sided writing surface of Thermo Scientific™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes.
  • Hinge design ensures specimen safety during processing and easy removal of cover while embedding
  • Base is compatible with metal lids and fits all popular size base molds (metal and disposable plastic)
  • Cassettes are conveniently available assembled or unassembled to facilitate usage in a labeling system or with metal lids
  • Solvent- and decalcifier-resistant and compatible with microwave techniques
  • Different colors allow for easy specimen coding
Specifications
Specifications
Biopsy Cassettes, Small Square Pore