Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human RIP2 Synthetic Peptide
Synthetic Peptide
Supplier: Invitrogen™ PEP0058
Description
RICK Synthetic Peptide, PEP-0058, from Invitrogen.
This peptide corresponds to 15 amino acids near the carboxy terminus of human RICK.PEP-0058 can be used as a blocking peptide with polyclonal antibody PA5-19933.Specifications
8767 | |
purified | |
Human RIP2 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
O43353 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
RIPK2 | |
0.02% sodium azide | |
50 μg | |
PBS |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction