Learn More
Invitrogen™ RICTOR Monoclonal Antibody (7B3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515681
Description
MA5-15681 targets RICTOR in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human, mouse, and Non-human primate samples. The MA5-15681 immunogen is purified recombinant fragment of human RICTOR expressed in E. Coli. MA5-15681 detects RICTOR which has a predicted molecular weight of approximately 192kDa.
RICTOR and MTOR (FRAP1) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
Specifications
RICTOR | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
Q6QI06, Q6R327 | |
Rictor | |
Purified recombinant fragment of human RICTOR expressed in E. Coli. | |
RUO | |
253260, 78757 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
7B3 | |
Unconjugated | |
Rictor | |
4921505C17Rik; 6030405M08Rik; AVO3; AVO3 homolog; AW492497; D530039E11Rik; DKFZp686B11164; Fyb; FYN binding protein; hAVO3; Kiaa1999; mAVO3; MGC39830; PIA; pianissimo; protein pianissimo; Rapamycin-insensitive companion of mTOR; rictor; RPTOR independent companion of MTOR complex 2; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Monkey | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.