Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIF1, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A300568AM

 View more versions of this product

Catalog No. 50-155-7939


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

May have a role in sperm development and fertilization.
Specifications

Specifications

RIF1
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
Q5UIP0
RAP1 interacting factor homolog, yeast, DKFZp434D1026, DKFZp781N1478, FLJ12870, rap1-interacting factor 1 homolog, telomere-associated protein RIF1
Rabbit
IgG
100 μl
4° C
Primary
55183
Western Blot
Unconjugated
RIF1
liquid
RIF1
Between 1425 and 1450
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit