Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rifampin, USP, 95-103%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
13292-46-1
C43H58N4O12
822.95
FZYOVNIOYYPUPY-UHFFFAOYNA-N
2,15,17,29-tetrahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-26-{[(4-methylpiperazin-1-yl)amino]methylidene}-6,23,27-trioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29)-heptaen-13-yl acetate
COC1C=COC2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)C(C)=CC=CC(C)C(O)C(C)C(O)C(C)C(OC(C)=O)C1C)C(=CNN1CCN(C)CC1)C(=O)C4=C3C2=O
13292-46-1
C43H58N4O12
822.95
FZYOVNIOYYPUPY-UHFFFAOYNA-N
2,15,17,29-tetrahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-26-{[(4-methylpiperazin-1-yl)amino]methylidene}-6,23,27-trioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29)-heptaen-13-yl acetate
COC1C=COC2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)C(C)=CC=CC(C)C(O)C(C)C(O)C(C)C(OC(C)=O)C1C)C(=CNN1CCN(C)CC1)C(=O)C4=C3C2=O

R1512, 13292-46-1, C43H58N4O12

Spectrum™ Rifampin, USP is an antibiotic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Crystallinity To pass test
Loss on Drying 0.02
Packaging Amber Glass Bottle
Related Substances To pass test
Percent Purity 95 to 103%
Grade USP
pH 4.5 to 6.5
Quantity 100 g