Learn More
Selleck Chemical LLCSupplier Diversity Partner Rifapentine 50mg 61379-65-5 MDL473  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S176050MG
Rifapentine (MDL473) is an antibiotic, which inhibits DNA-dependent RNA polymerase activity, used to treat tuberculosis.
Catalog No. 50-613-6