Learn More
Honeywell Rig Dog™ Cold Protect Gloves
Provides improved impact protection, comfort, and an enhanced grip for handling oily surfaces. The EN 511 winter lining is suitable for cold environments down to -20°F.
$42.00 - $43.00
Specifications
Hand Specific
Cut Resistance Rating ANSI A6
Material TPR
Certifications/Compliance ANSI/ISEA 138 Level 2
Series Rig Dog™ Cold Protect
View More Specs
Products 6
Catalog Number Mfr. No. Style Size Price Quantity & Availability  
Catalog Number Mfr. No. Style Size Price Quantity & Availability  
19-085-643
View Documents
Honeywell
43622BY/9L
Hook and Loop Closure Large
Pair for $42.00
Case of 36 PR for $1,523.34
Add to cart
19-085-644
View Documents
Honeywell
43622BY/10XL
Hook and Loop Closure X-Large
Pair for $42.00
Case of 36 PR for $1,523.34
Add to cart
19-085-645
View Documents
Honeywell
43622BY/11XXL
Hook and Loop Closure 2X-Large
Pair for $42.00
Case of 36 PR for $1,523.34
Add to cart
19-085-646
View Documents
Honeywell
43612BY/9L
Slip-On Cuff Large
Pair for $43.00
Case of 36 PR for $1,559.61
Add to cart
19-085-647
View Documents
Honeywell
43612BY/10XL
Slip-On Cuff X-Large
Pair for $43.00
Case of 36 PR for $1,559.61
Add to cart
19-085-648
View Documents
Honeywell
43612BY/11XXL
Slip-On Cuff 2X-Large
Pair for $43.00
Case of 36 PR for $1,559.61
Add to cart
Description
Description
  • Highly breathable, designed with an emphasis on wearer comfort
  • EN 511 winter lining for cold environments down to -20°F
  • Inner layer offers moisture absorption and a quick dry function to quickly absorb sweat and keep hands dry
  • Enhanced, level 2 impact protection from ANSI/ISEA 138
  • Additional layers provide A6/F level cut resistance, and guard against abrasions
  • Ergonomic design and flex-grooves for increased dexterity
  • Available in sizes: X-Small (6) to 2X-Large (11)
Specifications
Specifications
Specific
TPR
Rig Dog™ Cold Protect
Reinforced
Thermal Lined Gloves
ANSI A6
ANSI/ISEA 138 Level 2
Thermal
Cold Environments Down to -20°F
Individual Polybag