Learn More
ORS Nasco Rigid™ Heavy-Duty Bolt Cutters
One-step internal cam mechanism provides the quickest and simplest way to keep the blades precisely aligned
Manufacturer: ORS Nasco 63214228
Description
- Slim heads permit cutting close to surfaces
- Hardened jaws manufactured to assure the longest possible blade
- Handles and grips designed to allow greater mechanical advantage for crisp, controlled cutting
- Will cut through cutting rod, wire mesh, concrete form ties, rebar
Specifications
30 in. | |
Alloy Steel | |
9.98 lb. |