Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rilmenidine hemifumarate, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

207572-68-7
C24H36N4O6
476.574
LZFATBMLSYHRTC-WXXKFALUSA-N
2-[N-(Dicyclopropylmethyl)amino]oxazoline hemifumarate
45073450
(E)-but-2-enedioic acid;N-(dicyclopropylmethyl)-4,5-dihydro-1,3-oxazol-2-amine
C1CC1C(C2CC2)NC3=NCCO3.C1CC1C(C2CC2)NC3=NCCO3.C(=CC(=O)O)C(=O)O

α2 agonist. Also I1 ligand

I1-imidazoline binding site selective ligand and α2-adrenoceptor agonist. Possesses greater I1 vs α2 selectivity than the prototypical compound, clonidine. Also thought to enhance autophagy; shown to increase LC3-II levels in PC12 cells.
Quantity 50mg
Formula Weight 238.29
Chemical Name or Material Rilmenidine hemifumarate
Grade >99%

Recommended Storage : Store at -20°C