Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIMBP3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548812

Catalog No. PA548812

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

RIMBP3
Polyclonal
PBS with 0.09% sodium azide
Q9UFD9
RIMS-binding protein 3A, RIM-BP3A, RIMS-binding protein 31, RIM-BP31, RIMBP3, KIAA1666, RIMBP3A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
85376
Western Blot
Unconjugated
RIMBP3
Liquid
RIMBP3
KLH conjugated synthetic peptide between 180-209 amino acids from the N-terminal region of human RIMBP3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit