Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rimcazole dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Antagonist at σ receptors (IC50 values are 1480 and 386 nM at σ1 and σ2 receptors respectively). Also binds to the dopamine transporter (IC50 = 57.6 nM) and inhibits dopamine uptake. Reduces the stimulatory effects of cocaine in vivo.
Specifications
Quantity | 10 mg |
Formula Weight | 394.39 |
Percent Purity | >98% |
Chemical Name or Material | Rimcazole dihydrochloride |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction