Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIMKB Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309352100UL
Description
RIMKB Polyclonal specifically detects RIMKB in Mouse samples. It is validated for Western Blot.Specifications
RIMKB | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Beta-citrylglutamate synthase B, FAM80B, family with sequence similarity 80, member B, KIAA1238N-acetylaspartyl-glutamate synthetase, NAAG synthetase B, NAAGSEC 6.3.2.n6, ribosomal modification protein rimK-like family member B, Ribosomal protein S6 modification-like protein B | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Mouse RIMKB (NP_081940). Peptide sequence FRAVVMDEMVLTVEQGNLGLRISGELISAYPQVVVVRVPTPWVQSDSDIT | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
57494 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction