Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RIMS4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599707
Description
Antibody detects endogenous levels of total Rim4.
Regulates synaptic membrane exocytosis. Also a member of a family of synaptic proteins involved in neurotransmitter release.
Specifications
RIMS4 | |
Polyclonal | |
Unconjugated | |
Rims4 | |
6530406L02; C20orf190; rab3-interacting molecule 4; regulating synaptic membrane exocytosis 4; regulating synaptic membrane exocytosis protein 4; RIM 4; RIM4; RIM-4; RIM4 gamma; RIM4gamma; RIM4-gamma; RIMS4; synaptic regulatory protein RIM4gamma | |
Rabbit | |
Affinity chromatography | |
RUO | |
140730, 266976 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8CIX1, Q9H426 | |
Rims4 | |
A synthesized peptide derived from human RIMS4(Accession Q9H426), corresponding to amino acid residues A32-Y82. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction