Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIM4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549568
Description
Regulates synaptic membrane exocytosis.Specifications
RIM4 | |
Polyclonal | |
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9H426, P60191, Q8CIX1 | |
RIMS4 | |
synthetic peptide corresponding to 49-78aa of human RIMS4 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
RIMS4 | |
Regulating synaptic membrane exocytosis protein 4, RIM4 gamma, Rab3-interacting molecule 4, RIM 4, RIMS4, C20orf190 | |
Rabbit | |
Affinity chromatography | |
RUO | |
140730, 241770, 266976 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction