Learn More
RING1 Mouse anti-Human, Clone: OTI6F11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809256
Catalog No. 50-166-9375
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene belongs to the RING finger family, members of which encode proteins characterized by a RING domain, a zinc-binding motif related to the zinc finger domain. The gene product can bind DNA and can act as a transcriptional repressor. It is associated with the multimeric polycomb group protein complex. The gene product interacts with the polycomb group proteins BMI1, EDR1, and CBX4, and colocalizes with these proteins in large nuclear domains. It interacts with the CBX4 protein via its glycine-rich C-terminal domain. The gene maps to the HLA class II region, where it is contiguous with the RING finger genes FABGL and HKE4.
Specifications
Specifications
RING1
Monoclonal
Unconjugated
RING1
Lyophilized
RING1
Human recombit protein fragment corresponding to amino acids 88-362 of human RING1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI6F11
PBS with 8% trehalose and no preservative; pH 7.3
Q06587
RING1A, RNF1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6015