Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RING2/RNF2 Mouse anti-Human,Monkey,Mouse, Unconjugated, Clone: 3-3, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen RING2/RNF2
Applications ChIP assay, Immunocytochemistry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE18741MI
View Documents
MilliporeSigma
MABE1874100UG
100 μg
Each for $372.00
Add to cart
 
MABE18742MI
View Documents
MilliporeSigma
MABE187425UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

RING2/RNF2 Monoclonal antibody specifically detects RING2/RNF2 in Human,Monkey,Mouse samples. It is validated for Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

RING2/RNF2
Unconjugated
Mouse
E3 ubiquitin-protein ligase RING2;EC:2.3.2.27;RING finger protein 1B;RING finger protein 2;RING-type E3 ubiquitin transferase RING2
GST-tagged full length murine RING2 protein.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ChIP assay, Immunocytochemistry, Immunoprecipitation, Western Blot
Purified
Human, Monkey, Mouse
Rnf2, DinG, Ring1b
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit