Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ringers Rope Rescue Gloves

Encompass
Click to view available options
Color:
Black
Red
Size:
Small
Medium
Large
X-Large
2X-Large

Suited for all rope handling applications

Color Black
Material Neoprene, Synthetic Leather
Product Type Extrication Gloves
Size Large
For Use With (Application) Rescue, Rappelling, and SWAT Situations
Glove Cuff Style Hook and Loop