Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ringers Rope Rescue Gloves

Click to view available options
Color:
Black
Red
Size:
Small
Medium
Large
X-Large
2X-Large
Specifications
Color | Red |
Material | Neoprene, Synthetic Leather |
Product Type | Extrication Gloves |
Size | Large |
For Use With (Application) | Rescue, Rappelling, and SWAT Situations |
Glove Cuff Style | Hook and Loop |