Ringers™ SplitFit Air Rescue Gloves

Guard against cold, nicks, cuts, and bruises

Manufacturer: Ringers Gloves 14308

 View more versions of this product

Catalog No. 19-130-4280

  Check Availability
Add to Cart

Alert

Not intended for extrication or firefighting use.


    Specifications

    Cleaning Procedure Machine washable
    Glove Cuff Style Elastic
    For Use With (Application) guard against cold, nicks, cuts and bruises
    Size Small
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit