Learn More
Rink amide benzhydrylamine polymer resin, 1% crosslinked with DVB, 0.5mmol/g, ACROS Organics™
Manufacturer:  Acros Organics 349290050
 View more versions of this product
Catalog No. AC349290050
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
for solid support synthesis
Specifications
Specifications
crosslinking: 1% DVB
0.5 to 0.9mmol/g
Glass bottle
Brown-Pink to White
Beads
Rink amide benzhydrylamine polymer resin
200 to 400 mesh
5g
MFCD00677979