Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIOK3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259129

Catalog No. NBP259129


Add to cart

Description

Description

RIOK3 Polyclonal antibody specifically detects RIOK3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

RIOK3
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
DKFZp779L1370, EC 2.7.11.1, homolog of the Aspergillus nidulans sudD gene product, RIO kinase 3, RIO kinase 3 (yeast), serine/threonine-protein kinase RIO3, sudD (suppressor of bimD6, Aspergillus nidulans) homolog, sudD homolog, sudD suppressor of bimD6 homolog, SUDDsudD suppressor of Aspergillus nidulans bimD6 homolog
Rabbit
IgG
100 ul
Cell Cycle and Replication
Primary
8780
Western Blot, Immunoprecipitation
Western Blot, Immunoprecipitation
Affinity Purified
RIOK3
Maps to a region between 50 and 100
Immunogen affinity purified
RUO
Store at 4C.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only