Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIP1 Mouse anti-Canine, Human, Clone: OTI2G3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526427

Catalog No. PIMA526427


Add to cart

Description

Description

Serine-threonine kinase which transduces inflammatory and cell-death signals (programmed necrosis) following death receptors ligation, activation of pathogen recognition receptors (PRRs), and DNA damage. Upon activation of TNFR1 by the TNF-alpha family cytokines, TRADD and TRAF2 are recruited to the receptor. Phosphorylates DAB2IP at  ft.Ser-728 ft. in a TNF-alpha-dependent manner, and thereby activates the MAP3K5-JNK apoptotic cascade. Ubiquitination by TRAF2 via  ft.Lys-63 ft.-link chains acts as a critical enhancer of communication with downstream signal transducers in the mitogen-activated protein kinase pathway and the NF-kappa-B pathway, which in turn mediate downstream events including the activation of genes encoding inflammatory molecules. Polyubiquitinated protein binds to IKBKG/NEMO, the regulatory subunit of the IKK complex, a critical event for NF-kappa-B activation. Interaction with other cellular RHIM-containing adapters initiates gene activation and cell death. RIPK1 and RIPK3 association, in particular, forms a necrosis-inducing complex.
Specifications

Specifications

RIP1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q13546
OTTHUMP00000015955; RIP-1; cell death protein RIP; receptor interacting protein; receptor-interacting protein 1; receptor-interacting serine/threonine-protein kinase 1; serine/threonine-protein kinase RIP
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
102152063, 8737
Immunohistochemistry (Paraffin), Western Blot
OTI2G3
Unconjugated
RIPK1
Liquid
RIPK1
Human recombit protein fragment corresponding to amino acids 133-422 of RIPK1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit