Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ RIPA buffer with EGTA

Manufacturer:  Alfa Aesar™ J61951AK

 View more versions of this product

Catalog No. AAJ61951AK


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

Applications
RIPA buffer with EGTA is used in general cell-tissue lysis and subcellular orgnalle or molecular fractionation, including radio Immunoprecipitation. It is used for extracting proteins from cells. It is used for nuclear membrane disruption for nuclear extracts. It helps in disrupting protein-protein interactions.

Solubility
Fully miscible in water.

Notes
50mM Tris-HCl (pH 7.4), 150mM NaCl 1% NP-40, 0.5% sodium deoxycholate and 0.1% SDS with 1mM EGTA.
Specifications

Specifications

RIPA buffer with EGTA
Liquid
250mL
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit