Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIPK1 Rabbit anti-Human, Unconjugated, Clone: IGF-18, Abnova
Rabbit Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB20676
Description
RIPK1 Monoclonal antibody specifically detects RIPK1 in Human samples. It is validated for WB-Ce, Flow Cyt.Specifications
RIPK1 | |
IGF-18 | |
receptor (TNFRSF)-interacting serine-threonine kinase 1 | |
Liquid | |
Rabbit | |
IgG | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
Monoclonal | |
Human |
Flow Cytometry, Western Blot | |
Unconjugated | |
RIPK1 | |
FLJ39204, RIP, RIP1 | |
A synthetic peptide corresponding to human RIPK1 | |
100 μg | |
Primary | |
8737 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title