Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIPK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 201021AP150UL
Description
This antibody is specific to DSTYK.
This gene encodes a dual serine/threonine and tyrosine protein kinase which is expressed in multiple tissues. It is thought to function as a regulator of cell death. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
RIPK5 | |
Polyclonal | |
Unconjugated | |
DSTYK | |
DSTYK, Dusty PK, Dusty protein kinase, DustyPK, HDCMD38P, KIAA0472, RIP homologous kinase, RIP5, RIPK5, SgK496, Sugen kinase 496 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
213452, 25778, 304791 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6XUX1, Q6XUX2, Q6XUX3 | |
Dstyk | |
RIPK5 Fusion Protein Ag13678 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction