Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15484520UL

 View more versions of this product

Catalog No. NBP15484520


Add to cart

Description

Description

RIPK5 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

RIPK5
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
DSTYK
Synthetic peptides corresponding to RIPK5(receptor interacting protein kinase 5) The peptide sequence was selected from the middle region of RIPK5. Peptide sequence EECWQLMEACWDGDPLKRPLLGIVQPMLQGIMNRLCKSNSEQPNRGLDDS.
100 kDa
20ul
Protein Kinase
Primary
25778
Western Blot
Western Blot 1:100-1:2000
Q6XUX3-2
dual serine/threonine and tyrosine protein kinase, Dusty PK, Dusty protein kinase, DUSTYPK, KIAA0472RIP-homologous kinase, receptor interacting protein 5, receptor interacting protein kinase 5, Receptor-interacting serine/threonine-protein kinase 5, RIP5EC 2.7.12.1, RIPK5, sgK496, Sugen kinase 496
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit