Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals RITA, p53 activator III
Click to view available options
Quantity:
1 mg
10 mg
Description
A cell-permeable, p53-targeting, tricyclic thiophene derivative
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 mg |
Content And Storage | 2°C to 8°C |
For Use With (Application) | A cell-permeable, p53-targeting, tricyclic thiophene derivative |
Formula Weight | 292.40g/mol |
Sensitivity | Light Sensitive |
Product Type | Signal Transduction Reagent |
Form | Crystalline solid |
Safety and Handling
Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction