Learn More
Alfa Aesar™ RITA, p53 activator III
A cell-permeable, p53-targeting, tricyclic thiophene derivative
Manufacturer:  Alfa Aesar™ J65379MA
 View more versions of this product
Catalog No. AAJ65379MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
10mg
Crystalline Solid
213261-59-7
292.40g/mol
Light Sensitive
2° to 8°C
A cell-permeable, p53-targeting, tricyclic thiophene derivative
C14H12O3S2
5,5′-(2,5-Furandiyl)bis-2-thiophenemethanol; NSC 652287
Signal Transduction Reagent