Learn More
Epredia™ Rite-On Frosted Thick Slide
Label slides quickly and easily with Epredia™ Gold Seal™ Rite-On™ and  Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass.
Manufacturer:  Epredia™ 3051002
 View more versions of this product
Catalog No. 12-518-103
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
All finished slides are sealed with a desiccant in an air tight, lint-free, hermetically sealed Moistur-Barrier™ package, keeping slides dry during shipping and storage. Slides work well in hematology, histology, cytology, microbiology and urinalysis; as well as other laboratory and research applications.
  • Smooth marking area for effortless writing, automated printing, or printed labels
  • Each case features an inner barrier-bag sealed with desiccant packs to protect from moisture contamination
  • Slide boxes convert into convenient slide storage files
  • Reduced iron content for low self-fluorescence
  • Manufactured from pure, water-white glass
  • Precleaned for immediate use
  • Part No. 3032-002 is used for fluorescent antibody examination
Specifications
Specifications
76 mm
1 mm
Yes
Microscope Slide
White
Rite-On Frosted Slide
Packed 72 slides per box, 50 boxes per case
3 x 1 in.
Ground
Glass
White Epoxy Ink Frosted End, 1 Side
25 mm
90°
1 in.
1 in.