Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Rite-On Frosted Thick Slide
Label slides quickly and easily with Epredia™ Gold Seal™ Rite-On™ and Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass.
Supplier: Epredia™ 3032002
Description
All finished slides are sealed with a desiccant in an air tight, lint-free, hermetically sealed Moistur-Barrier™ package, keeping slides dry during shipping and storage. Slides work well in hematology, histology, cytology, microbiology and urinalysis; as well as other laboratory and research applications.- Smooth marking area for effortless writing, automated printing, or printed labels
- Each case features an inner barrier-bag sealed with desiccant packs to protect from moisture contamination
- Slide boxes convert into convenient slide storage files
- Reduced iron content for low self-fluorescence
- Manufactured from pure, water-white glass
- Precleaned for immediate use
- Part No. 3032-002 is used for fluorescent antibody examination
Specifications
76 mm | |
1 mm | |
White Epoxy Ink Frosted End, 1 Side, with two 10mm diameter circles | |
Microscope Slide | |
White | |
Rite-On Frosted Fluorescent Antibody Slide | |
Packed 72 slides per box, 50 boxes per case | |
3 x 1 in. (75 x 25 mm) |
Ground | |
Glass | |
25 mm | |
Yes | |
90° | |
3 in. | |
1 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction