Learn More
Epredia™ Rite-On Frosted Thick Slide
Label slides quickly and easily with Epredia™ Gold Seal™ Rite-On™ and  Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass.
Manufacturer:  Epredia™ 3031002
 View more versions of this product
Catalog No. 12-569-20
Add to cart
Description
Description
All finished slides are sealed with a desiccant in an air tight, lint-free, hermetically sealed Moistur-Barrier™ package, keeping slides dry during shipping and storage. Slides work well in hematology, histology, cytology, microbiology and urinalysis; as well as other laboratory and research applications.
  • Smooth marking area for effortless writing, automated printing, or printed labels
  • Each case features an inner barrier-bag sealed with desiccant packs to protect from moisture contamination
  • Slide boxes convert into convenient slide storage files
  • Reduced iron content for low self-fluorescence
  • Manufactured from pure, water-white glass
  • Precleaned for immediate use
  • Part No. 3032-002 is used for fluorescent antibody examination
Specifications
Specifications
Cytology Slide
Yes
Ground
76mm
White
90°
3 in.
3 x 1 in. (76 x 25mm)
Glass
Yes
1mm
25mm
Fully Sandblast Frosted, One Side
Rite-On Fully Frosted Slide
Packed 72 slides per box, 50 boxes per case
1 in.