Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rituximab, Rabbit anti-Many, Alexa Fluor 700, Clone: 2260A, R&D Systems™

Rabbit Monoclonal Antibody

Manufacturer:  R&D Systems FAB9630N100UG

Catalog No. FAB9630N100


Add to cart
This item is not returnable. View return policy

Description

Description

Rituximab Monoclonal antibody specifically detects Rituximab in Multi-Species Samples. It is validated for Flow Cytometry.
Specifications

Specifications

Rituximab
2260A
Flow Cytometry 0.25-1 ug/10^6 cells
Purified
Rabbit
IgG
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Many
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
B lymphocyte antigen CD20, B lymphocyte surface antigen B1, Bp35, CD20, LEU 16, LEU16, Membrane spanning 4 domains subfamily A member 1, MS4A1
Full-length Rituximab
Protein A or G purified from cell culture supernatant
RUO
Primary
Detects Rituximab in Western blots in direct ELISAs. 100% cross-reactivity with Tositumomab and Ibritumomab (95% or higher sequence identity with Rituximab) and no cross-reactivity with Cetuximab, Trastuzumab, Brentuximab, Gemtuzumab, Adalimumab, Solanezumab, Teprotumumab, and Aducanumab is observed.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit