Learn More
Selleck Chemical LLCSupplier Diversity Partner Rivaroxaban 10mg 366789-02-8 BAY 59-7939  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S300210MG
Rivaroxaban (BAY 59-7939) is a direct inhibitor of Factor Xa with Ki and IC50 of 0.4 nM and 0.7 nM in cell-free assays, respectively. It is selective for human factor Xa, for which it has >10 000-fold greater selectivity than for other biologically relevant serine proteases (IC50 >20 μM).
Catalog No. 50-618-7