Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rizatriptan benzoate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
5-HT1B/1D agonist. Antimigraine agent; attenuates effects of CGRP on light aversion in mice. Orally bioavailable.
Specifications
Color | White |
Quantity | 10 mg |
Formula Weight | 391.47 |
Percent Purity | >99% |
Physical Form | Solid |
Chemical Name or Material | Rizatriptan benzoate |
Safety and Handling
Recommended Storage | Store at RT |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction