Rizatriptan benzoate, Tocris™

5-HT1B/1D agonist. Antimigraine agent; attenuates effects of CGRP on light aversion in mice. Orally bioavailable

Manufacturer: R&D Systems 5136/50

Quantity 50mg

 View more versions of this product

Catalog No. 51-365-0R

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 145202-66-0
    Physical Form Solid
    Assay Percent Range >99%
    Molecular Formula C15H19N5 · C7H6O2
    Formula Weight 391.47
    Chemical Name or Material Rizatriptan benzoate

    Specifications

    Physical Form Solid
    Formula Weight 391.47
    Color White
    Quantity 50mg
    Assay Percent Range >99%
    Molecular Formula C15H19N5 · C7H6O2
    CAS 145202-66-0
    Recommended Storage Store at RT
    Chemical Name or Material Rizatriptan benzoate

    Store at RT

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit