Learn More
Pyramex™ RJ32 Series Winter Wear Bomber Jacket
SDP
Pyramex™ Safety Hi -Vis Winter Wear
$115.40
Specifications
Material Weatherproof Polyester
Coating Polyurethane
Liner Material Quilted Polyester Cotton
Color Hi-vis Lime
Tape Type China Brand Flat Silver Reflective Tape
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-555-270
SDP
View Documents
Pyramex™
RJ3210M
Medium
Each for $115.40
Add to cart
19-555-269
SDP
View Documents
Pyramex™
RJ3210L
Large
Each for $115.40
Add to cart
19-555-276
SDP
View Documents
Pyramex™
RJ3210XL
X-Large
Each for $115.40
Add to cart
19-555-271
SDP
View Documents
Pyramex™
RJ3210X2
2X-Large
Each for $115.40
Add to cart
19-555-272
SDP
View Documents
Pyramex™
RJ3210X3
3X-Large
Each for $115.40
Add to cart
19-555-273
SDP
View Documents
Pyramex™
RJ3210X4
4X-Large
Each for $115.40
Add to cart
19-555-274
SDP
View Documents
Pyramex™
RJ3210X5
5X-Large
Each for $115.40
Add to cart
19-555-275
SDP
View Documents
Pyramex™
RJ3210X6
6X-Large
Each for $115.40
Add to cart
Description
Description
  • Weatherproof polyester shell with quilted lining
  • Weatherproof elastic band at the waist and wrists
  • 2 in. silver reflective material
  • Concealed hood
  • Zipper front closure
  • 5 pockets: 4 outside, 1 inside
  • Cell phone/radio chest pocket with clear panel on outside shell
  • Mic tabs and pencil pocket
Specifications
Specifications
Weatherproof Polyester
Quilted Polyester Cotton
China Brand Flat Silver Reflective Tape
Concealed, Detachable
5
Polyurethane
Hi-vis Lime
Nylon Zipper
Elastic
ANSI/ISEA 107-2015; Type R - Class 3