Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyramex™ RJR33 Series Winter Wear Jacket

Pyramex™ Safety Hi -Vis Winter Wear
$88.42
Specifications
Material | Polyester |
---|---|
Color | Hi-vis Lime, Black |
Closure Type | Nylon Zipper |
Certifications/Compliance | ANSI/ISEA 107-2015; Type R - Class 3 with sleeves hi-vis side only; Type R - Class 2 without sleeves hi-vis side only |
Coating | Water repellent treatment (hi-vis side only); PU white coating (black side only) |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-555-281
![]() |
Pyramex™
RJR3310X4 |
4X-Large |
Each for $88.42
|
|
19-555-278
![]() |
Pyramex™
RJR3310M |
Medium | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
19-555-277
![]() |
Pyramex™
RJR3310L |
Large | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
19-555-283
![]() |
Pyramex™
RJR3310XL |
X-Large | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
19-555-279
![]() |
Pyramex™
RJR3310X2 |
2X-Large | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
19-555-280
![]() |
Pyramex™
RJR3310X3 |
3X-Large | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
19-555-282
![]() |
Pyramex™
RJR3310X5 |
5X-Large | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Water repellent 4-in-1 reversible quilted polyfil jacket
- Removable, zip-out sleeves – converts Class 3 jacket to Class 2 vest
- Reverses from hi-vis lime/black bottom to all black
- Silver reflective tape on hi-vis side
Specifications
Polyester | |
Nylon Zipper | |
Water repellent treatment (hi-vis side only); PU white coating (black side only) | |
3 per side | |
China Brand Silver Reflective Stripes |
Hi-vis Lime, Black | |
ANSI/ISEA 107-2015; Type R - Class 3 with sleeves hi-vis side only; Type R - Class 2 without sleeves hi-vis side only | |
Polyester | |
Removable, Zip-out |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title