Learn More
Pyramex™ RJR34 Series Winter Wear Windbreaker
SDP
Pyramex™ Safety Hi -Vis Winter Wear
$111.00
Specifications
Material Polyester
Color Hi-vis Lime
Closure Type Nylon Zipper
Certifications/Compliance Meets ANSI/ISEA 107-2015; Type R - Class 3 with sleeves hi-vis side only; Type R - Class 2 without sleeves hi-vis side only
Coating TPU Coating and Water Resistant Treatment
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-555-285
SDP
View Documents
Pyramex™
RJR3410M
Medium
Each for $111.00
Add to cart
19-555-284
SDP
View Documents
Pyramex™
RJR3410L
Large
Each for $111.00
Add to cart
19-555-290
SDP
View Documents
Pyramex™
RJR3410XL
X-Large
Each for $111.00
Add to cart
19-555-286
SDP
View Documents
Pyramex™
RJR3410X2
2X-Large
Each for $111.00
Add to cart
19-555-287
SDP
View Documents
Pyramex™
RJR3410X3
3X-Large
Each for $111.00
Add to cart
19-555-288
SDP
View Documents
Pyramex™
RJR3410X4
4X-Large
Each for $111.00
Add to cart
19-555-289
SDP
View Documents
Pyramex™
RJR3410X5
5X-Large
Each for $111.00
Add to cart
Description
Description
  • Water resistant 4-in-1 reversible windbreaker polyester jacket
  • Weatherproof elastic band at wrists
  • Removable zip-out sleeves — converts Class 3 jacket to Class 2 vest
  • Reverses from hi-vis lime to gray/black
  • Silver reflective tape on hi-vis side
  • Hi-vis reflective piping on gray/black side
  • Bottom hem adjustment — bungee cord/toggle pull
  • Mic tab on hi-vis side
Specifications
Specifications
Polyester
Nylon Zipper
TPU Coating and Water Resistant Treatment
Removable, Zip-out
Hi-vis Lime
Meets ANSI/ISEA 107-2015; Type R - Class 3 with sleeves hi-vis side only; Type R - Class 2 without sleeves hi-vis side only
3 per side
China Brand Silver Reflective Stripes