Learn More
RKI Sample Draw for GX-82 and GX-82-A GasTech Gas Monitors
Accessory for GX-82 and GX-82-A GasTech Gas Monitors.
Manufacturer:  RKI 811157RK
Catalog No. 19-024200
Add to cart
Description
Description
  • Hand-operated
  • With 10 ft. hose and probe
  • Not for use with SO2 or CI2
Specifications
Specifications
GX-82 and GX-82-A GasTech Gas Monitors
10 ft. hose and probe
Hand-operated sample draw