RKI Diffusion Sensor/Transmitters
Manufacturer: RKI InstrumentsSupplier Diversity Partner GDK8ACL2
 View more versions of this product
Catalog No. 18-999-1787
  Check Availability
Add to Cart
    Specifications
    Product Type Diffusion sensor/transmitter
    Concentration 0-3.0ppm
    Detects Chlorine (Cl2)
  • Description & Specifications
Related Products
Product Content Feedback