RKI™ Eagle Series Model Sensors
Manufacturer: RKI InstrumentsSupplier Diversity Partner ES23AHPH3
 View more versions of this product
Catalog No. 18-999-1819
  Check Availability
Add to Cart
    Specifications
    Measurement Range 0 - 1.0ppm (no H2 response)
    Sensor Types Phosphine
    For Use With (Equipment) GD-K8A Diffusion, GD-K7D2 Sample Drawing Transmitter For Toxic Gases, EAGLE Portable Series, SC-90 Series
  • Description & Specifications
Related Products
Product Content Feedback