RKI™ Eagle Series Model Sensors
Manufacturer: RKI InstrumentsSupplier Diversity Partner ES23AHSIH4
 View more versions of this product
Catalog No. 18-999-1820
  Check Availability
Add to Cart
    Specifications
    Measurement Range 0 - 15ppm, old style (no H2 response)
    Sensor Types Silane
    For Use With (Equipment) GD-K8A Diffusion, GD-K7D2 Sample Drawing Transmitter For Toxic Gases, EAGLE Portable Series, SC-90 Series
  • Description & Specifications
Related Products
Product Content Feedback