Learn More
RKI Carbon Monoxide (CO) Sensor for RKI Gas Monitors
For use with GX82, GX86, GX-82A, GX-86A, GX-91B and GX-4000 portable gas monitors.
Manufacturer:  RKI 652003RK
Catalog No. 19-023756
Add to cart
Description
Description
  • Sensors are mounted in the diffusion head at the bottom of the unit
  • The diffusion grill protects the sensors; grill easily snaps off the instrument case so you can conveniently replace the sensors
  • Carbon Monoxide (CO) sensor is contained in a cylindrical metal shell, with a bayonet pin base for easy replacement
  • Shell forms one electrical contact, and a button on the base is the other contact
  • The perforations at the end of the shell allow atmosphere to diffuse into the sensor
Specifications
Specifications
GX82, GX86, GX-82A, GX-86A, GX-91B and GX-4000 portable gas monitors
Carbon Monoxide (CO) Sensor