Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RKIP/PBP Antibody (003), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290295AF750
Description
RKIP/PBP Monoclonal specifically detects RKIP/PBP in Human samples. It is validated for Western Blot, ELISA.Specifications
RKIP/PBP | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
3 | |
Western Blot, ELISA | |
HCNP, hippocampal cholinergic neurostimulating peptide, Neuropolypeptide h3, PBPPEBP-1, PEBPHCNPpp, phosphatidylethanolamine binding protein 1, phosphatidylethanolamine-binding protein 1, prostatic binding protein, Prostatic-binding protein, Raf kinase inhibitory protein, RKIPRaf kinase inhibitor protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human RKIP/PBP (NP_002558.1; Met1-Lys187). | |
0.1 mL | |
Cancer, Phospho Specific | |
5037 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction