Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLVS2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548193

Catalog No. PA548193

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Required for normal morphology of late endosomes and/or lysosomes in neurons. Binds phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2).
Specifications

Specifications

CLVS2
Polyclonal
PBS with 0.09% sodium azide
Q5SYC1, Q8BG92
Clavesin-2, Retinaldehyde-binding protein 1-like 2, CLVS2, C6orf212, C6orf213, RLBP1L2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
134829, 215890
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CLVS2
Liquid
CLVS2
KLH conjugated synthetic peptide between 251-279 amino acids from the C-terminal region of human RLBP1L2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit