Learn More
RLTPR Mouse anti-Human, Mouse, Alexa Fluor™ 350, Clone: EM-53, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP262215AF350
Catalog No. NB002602
Add to cart
Description
Description
RLTPR Monoclonal specifically detects RLTPR in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
RLTPR
EM-53
Western Blot, Flow Cytometry
Purified, Conjugated
RLTPR
Murine Rltpr (amino acids 1147-1397)
Protein A purified
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody EM-53 recognizes RLTPR/ CARMIL2, a protein playing a role in actin filament elongation.
Western Blot, Flow Cytometry
Alexa Fluor 350
50mM Sodium Borate with 0.05% Sodium Azide
CARMIL2, CARMIL2b, leucine rich repeat containing 16C, leucine-rich repeat-containing protein 16C, LRRC16C, RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing, RGD, leucine-rich repeat, tropomodulin and proline-rich containing protein, RGD, leucine-rich repeat, tropomodulin and proline-rich-containing protein
Mouse
IgG1 Kappa
0.1 mL
Primary
146206
Human, Mouse