Learn More
Akron Biotechnology LLCSupplier Diversity Partner RM IL-2 100UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK83230100
rM IL-2 is an immunomodulatory cytokine produced by stimulated T-cells. Its activity is signaled via interaction with the IL-2R receptor. 100ug
Catalog No. 50-753-3378